Assay Details
| Gene Symbol: tp53rk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020550.2 | NP_001018386.2 | 1 - 2 | 289 | 90 | |
| GenBank mRNA | BC095119.1 | - | 1 - 2 | 285 | 90 | |
| BC165061.1 | - | 1 - 2 | 266 | 90 | ||
| EH547402.1 | - | 1 - 2 | 238 | 90 | ||
More Information
Set Membership: |
|

tp53rk
Genomic Map