Assay Details
| Gene Symbol: rragca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020635.1 | NP_001018471.1 | 4 - 5 | 836 | 105 | |
| GenBank mRNA | BC095626.1 | - | 4 - 5 | 836 | 105 | |
| BC154154.1 | - | 4 - 5 | 706 | 105 | ||
More Information
Set Membership: |
|

rragca
Genomic Map