Assay Details
| Gene Symbol: clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020684.1 | NP_001018520.1 | 6 - 7 | 610 | 69 | |
| GenBank mRNA | BC095769.1 | - | 6 - 7 | 610 | 69 | |
More Information
Set Membership: |
|

clpp
Genomic Map