Assay Details
| Gene Symbol: spc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024440.1 | NP_001019611.1 | 5 - 6 | 544 | 74 | |
| GenBank mRNA | BC076486.1 | - | 4 - 5 | 525 | 74 | |
| BC095251.1 | - | 5 - 6 | 544 | 74 | ||
| BC152264.1 | - | 5 - 6 | 539 | 74 | ||
| BC164821.1 | - | 4 - 5 | 526 | 74 | ||
More Information
Set Membership: |
|

spc24
Genomic Map