Assay Details
| Gene Symbol: tsnax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030104.1 | NP_001025275.1 | 2 - 3 | 331 | 120 | |
| GenBank mRNA | BC097140.1 | - | 2 - 3 | 331 | 120 | |
More Information
Set Membership: |
|

tsnax
Genomic Map