Assay Details
| Gene Symbol: rab11fip1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003199114.5 | XP_003199162.3 | 1673 | 72 | ||
| XM_017357512.2 | XP_017213001.1 | 1667 | 72 | |||
| GenBank mRNA | BC093155.1 | - | 2 - 3 | 847 | 72 | |
| BC108064.1 | - | 867 | 72 | |||
More Information
Set Membership: |
|

rab11fip1a
Genomic Map