Assay Details
| Gene Symbol: sept4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037379.1 | NP_001032456.1 | 11 - 12 | 1606 | 59 | |
| GenBank mRNA | BC109428.1 | - | 11 - 12 | 1606 | 59 | |
More Information
Set Membership: |
|

sept4a
Genomic Map