Assay Details
| Gene Symbol: sept4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037379.1 | NP_001032456.1 | 7 - 8 | 1142 | 105 | |
| GenBank mRNA | BC109428.1 | - | 7 - 8 | 1142 | 105 | |
More Information
Set Membership: |
|

sept4a
Genomic Map