Assay Details
| Gene Symbol: slc6a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039972.1 | NP_001035061.1 | 1 - 2 | 469 | 63 | |
| XM_005157654.4 | XP_005157711.1 | 4 - 5 | 1647 | 63 | ||
| XM_005157655.4 | XP_005157712.1 | 4 - 5 | 1570 | 63 | ||
| XM_009291668.3 | XP_009289943.1 | 5 - 6 | 1839 | 63 | ||
| XM_009291669.3 | XP_009289944.1 | 5 - 6 | 1757 | 63 | ||
| XM_009291670.3 | XP_009289945.1 | 3 - 4 | 1378 | 63 | ||
| XM_009291671.3 | XP_009289946.1 | 5 - 6 | 1135 | 63 | ||
| XM_009291672.3 | XP_009289947.1 | 3 - 4 | 1296 | 63 | ||
| XM_009291673.3 | XP_009289948.1 | 1 - 2 | 208 | 63 | ||
| GenBank mRNA | BC163766.1 | - | 1 - 2 | 469 | 63 | |
| BC163777.1 | - | 1 - 2 | 469 | 63 | ||
| DQ285098.1 | - | 1 - 2 | 469 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009291673.3 transcripts.
Set Membership: |
|

slc6a4a
Genomic Map