Assay Details
| Gene Symbol: slc6a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039972.1 | NP_001035061.1 | 2 - 3 | 605 | 95 | |
| XM_005157654.4 | XP_005157711.1 | 5 - 6 | 1783 | 95 | ||
| XM_005157655.4 | XP_005157712.1 | 5 - 6 | 1706 | 95 | ||
| XM_009291668.3 | XP_009289943.1 | 6 - 7 | 1975 | 95 | ||
| XM_009291669.3 | XP_009289944.1 | 6 - 7 | 1893 | 95 | ||
| XM_009291670.3 | XP_009289945.1 | 4 - 5 | 1514 | 95 | ||
| XM_009291671.3 | XP_009289946.1 | 6 - 7 | 1271 | 95 | ||
| XM_009291672.3 | XP_009289947.1 | 4 - 5 | 1432 | 95 | ||
| XM_009291673.3 | XP_009289948.1 | 2 - 3 | 344 | 95 | ||
| GenBank mRNA | BC163766.1 | - | 2 - 3 | 605 | 95 | |
| BC163777.1 | - | 2 - 3 | 605 | 95 | ||
| DQ285098.1 | - | 2 - 3 | 605 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009291673.3 transcripts.
Set Membership: |
|

slc6a4a
Genomic Map