Assay Details
| Gene Symbol: f9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040310.1 | NP_001035400.1 | 5 - 6 | 553 | 135 | |
| GenBank mRNA | BC115275.1 | - | 5 - 6 | 553 | 135 | |
More Information
Set Membership: |
|

f9b
Genomic Map