Assay Details
| Gene Symbol: kcnh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042722.2 | NP_001036187.2 | 7 - 8 | 2413 | 90 | |
| XM_021480879.1 | XP_021336554.1 | 7 - 8 | 2388 | 90 | ||
| XM_021480881.1 | XP_021336556.1 | 3 - 4 | 1817 | 90 | ||
| GenBank mRNA | DQ022613.1 | - | 7 - 8 | 1953 | 90 | |
| HM209079.1 | - | 7 - 8 | 1953 | 90 | ||
More Information
Set Membership: |
|

kcnh2a
Genomic Map