Assay Details
| Gene Symbol: kcnh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042722.2 | NP_001036187.2 | 10 - 11 | 3065 | 119 | |
| XM_021480879.1 | XP_021336554.1 | 10 - 11 | 3040 | 119 | ||
| XM_021480881.1 | XP_021336556.1 | 6 - 7 | 2469 | 119 | ||
| GenBank mRNA | DQ022613.1 | - | 10 - 11 | 2605 | 119 | |
| HM209079.1 | - | 10 - 11 | 2605 | 119 | ||
More Information
Set Membership: |
|

kcnh2a
Genomic Map