Assay Details
| Gene Symbol: dusp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044307.1 | NP_001037772.1 | 2 - 3 | 344 | 125 | |
| GenBank mRNA | BC162702.1 | - | 2 - 3 | 395 | 125 | |
| BC162708.1 | - | 2 - 3 | 395 | 125 | ||
More Information
Set Membership: |
|

dusp3b
Genomic Map