Assay Details
| Gene Symbol: wnt2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044344.1 | NP_001037809.1 | 4 - 5 | 1181 | 79 | |
| GenBank mRNA | BC162931.1 | - | 4 - 5 | 1207 | 79 | |
| BC162939.1 | - | 4 - 5 | 1207 | 79 | ||
| DQ231559.1 | - | 4 - 5 | 1181 | 79 | ||
More Information
Set Membership: |
|

wnt2bb
Genomic Map