Assay Details
| Gene Symbol: ppp1r13bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044824.2 | NP_001038289.2 | 5 - 6 | 506 | 52 | |
| XM_005160501.4 | XP_005160558.1 | 4 - 5 | 508 | 52 | ||
| XM_009294640.3 | XP_009292915.1 | 4 - 5 | 509 | 52 | ||
| XM_009294641.3 | XP_009292916.1 | 4 - 5 | 510 | 52 | ||
| XM_009294642.3 | XP_009292917.1 | 5 - 6 | 759 | 52 | ||
| XM_009294643.3 | XP_009292918.1 | 2 - 3 | 177 | 52 | ||
| GenBank mRNA | BC139881.1 | - | 5 - 6 | 506 | 52 | |
More Information
Set Membership: |
|

ppp1r13bb
Genomic Map