Assay Details
| Gene Symbol: ppp1r13bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044824.2 | NP_001038289.2 | 9 - 10 | 1137 | 59 | |
| XM_005160501.4 | XP_005160558.1 | 8 - 9 | 1139 | 59 | ||
| XM_009294640.3 | XP_009292915.1 | 8 - 9 | 1140 | 59 | ||
| XM_009294642.3 | XP_009292917.1 | 9 - 10 | 1390 | 59 | ||
| XM_009294643.3 | XP_009292918.1 | 6 - 7 | 808 | 59 | ||
| GenBank mRNA | BC139881.1 | - | 9 - 10 | 1137 | 59 | |
More Information
Set Membership: |
|

ppp1r13bb
Genomic Map