Assay Details
| Gene Symbol: neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044909.2 | NP_001038374.1 | 2 - 3 | 331 | 122 | |
| XM_009294220.3 | XP_009292495.1 | 2 - 3 | 276 | 122 | ||
| GenBank mRNA | BC133832.1 | - | 2 - 3 | 331 | 122 | |
| LC373154.1 | - | 2 - 3 | 263 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009294220.3 transcripts.
Set Membership: |
|

neu1
Genomic Map