Assay Details
| Gene Symbol: rad21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045120.2 | NP_001038585.1 | 5 - 6 | 692 | 68 | |
| XM_005159782.4 | XP_005159839.1 | 6 - 7 | 844 | 68 | ||
More Information
Set Membership: |
|

rad21b
Genomic Map