Assay Details
| Gene Symbol: znf384b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045163.1 | NP_001038628.1 | 6 - 7 | 691 | 73 | |
| GenBank mRNA | BC129180.1 | - | 6 - 7 | 693 | 73 | |
More Information
Set Membership: |
|

znf384b
Genomic Map