Assay Details
| Gene Symbol: tmem14ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045438.1 | NP_001038903.1 | 3 - 4 | 249 | 79 | |
| XM_005162574.4 | XP_005162631.2 | 3 - 4 | 204 | 79 | ||
| XM_005162575.4 | XP_005162632.1 | 3 - 4 | 283 | 79 | ||
| GenBank mRNA | BC122294.1 | - | 3 - 4 | 249 | 79 | |
More Information
Set Membership: |
|

tmem14ca
Genomic Map