Assay Details
| Gene Symbol: zgc:153247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045442.1 | NP_001038907.1 | 8 - 9 | 1476 | 67 | |
| GenBank mRNA | BC122204.1 | - | 8 - 9 | 1476 | 67 | |
More Information
Set Membership: |
|

zgc:153247
Genomic Map