Assay Details
| Gene Symbol: rhebl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076748.2 | NP_001070216.1 | 6 - 7 | 692 | 135 | |
| GenBank mRNA | BC124281.1 | - | 6 - 7 | 598 | 135 | |
More Information
Set Membership: |
|

rhebl1
Genomic Map