Assay Details
| Gene Symbol: fbxo11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159668.1 | NP_001153140.1 | 15 - 16 | 1866 | 98 | |
| XM_005169554.4 | XP_005169611.1 | 15 - 16 | 2107 | 98 | ||
| GenBank mRNA | BC125857.1 | - | 15 - 16 | 1986 | 98 | |
More Information
Set Membership: |
|

fbxo11a
Genomic Map