Assay Details
| Gene Symbol: zgc:158316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080074.1 | NP_001073543.1 | 4 - 5 | 272 | 69 | |
| GenBank mRNA | BC128840.1 | - | 4 - 5 | 272 | 69 | |
| Gene Symbol: LOC100536464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021471062.1 | XP_021326737.1 | 3 - 4 | 531 | 69 | |
More Information
Set Membership: |
|

zgc:158316
Genomic Map