Assay Details
| Gene Symbol: zgc:158316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080074.1 | NP_001073543.1 | 6 - 7 | 529 | 68 | |
| GenBank mRNA | BC128840.1 | - | 6 - 7 | 529 | 68 | |
| Gene Symbol: LOC100536464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021471062.1 | XP_021326737.1 | 5 - 6 | 788 | 68 | |
More Information
Set Membership: |
|

zgc:158316
Genomic Map