Assay Details
| Gene Symbol: prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080562.2 | NP_001074031.2 | 4 - 5 | 792 | 65 | |
| GenBank mRNA | BC129320.1 | - | 4 - 5 | 772 | 65 | |
| EH506502.1 | - | 4 - 5 | 795 | 65 | ||
More Information
Set Membership: |
|

prss16
Genomic Map