Assay Details
| Gene Symbol: engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080578.2 | NP_001074047.2 | 11 - 12 | 1600 | 63 | |
| XM_021470912.1 | XP_021326587.1 | 11 - 12 | 1662 | 63 | ||
| XM_021470913.1 | XP_021326588.1 | 9 - 10 | 1269 | 63 | ||
| GenBank mRNA | BC129351.1 | - | 11 - 12 | 1602 | 63 | |
More Information
Set Membership: |
|

engase
Genomic Map