Assay Details
| Gene Symbol: spam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080685.1 | NP_001074154.1 | 3 - 4 | 1072 | 157 | |
| GenBank mRNA | BC129367.1 | - | 3 - 4 | 1072 | 157 | |
More Information
Set Membership: |
|

spam1
Genomic Map