Assay Details
| Gene Symbol: cacnb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083555.1 | NP_001077024.1 | 14 - 15 | 1436 | 145 | |
| XM_017353586.2 | XP_017209075.1 | 14 - 15 | 1613 | 145 | ||
| GenBank mRNA | DQ372946.1 | - | 14 - 15 | 1436 | 145 | |
More Information
Set Membership: |
|

cacnb3a
Genomic Map