Assay Details
| Gene Symbol: cacnb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083555.1 | NP_001077024.1 | 1 - 2 | 182 | 81 | |
| XM_005161994.4 | XP_005162051.1 | 1 - 2 | 353 | 81 | ||
| XM_005161995.4 | XP_005162052.1 | 1 - 2 | 353 | 81 | ||
| XM_017353586.2 | XP_017209075.1 | 1 - 2 | 353 | 81 | ||
| GenBank mRNA | BC162364.1 | - | 1 - 2 | 161 | 81 | |
| BC162649.1 | - | 1 - 2 | 161 | 81 | ||
| DQ372946.1 | - | 1 - 2 | 182 | 81 | ||
More Information
Set Membership: |
|

cacnb3a
Genomic Map