Assay Details
Gene Symbol: rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089434.1 | NP_001082903.1 | 1 - 2 | 158 | 66 | |
XM_009292345.3 | XP_009290620.1 | 1 - 2 | 301 | 66 | ||
GenBank mRNA | BC152599.1 | - | 1 - 2 | 142 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
