Assay Details
| Gene Symbol: tdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099417.1 | NP_001092887.1 | 2 - 3 | 387 | 70 | |
| GenBank mRNA | BC142842.1 | - | 2 - 3 | 387 | 70 | |
More Information
Set Membership: |
|

tdh2
Genomic Map