Assay Details
| Gene Symbol: zzz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100027.1 | NP_001093497.1 | 10 - 11 | 2470 | 57 | |
| XM_005163282.4 | XP_005163339.1 | 11 - 12 | 2774 | 57 | ||
More Information
Set Membership: |
|

zzz3
Genomic Map