Assay Details
| Gene Symbol: pparab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102567.1 | NP_001096037.1 | 5 - 6 | 885 | 67 | |
| XM_009297698.3 | XP_009295973.1 | 6 - 7 | 1273 | 67 | ||
| GenBank mRNA | DQ839548.1 | - | 5 - 6 | 885 | 67 | |
More Information
Set Membership: |
|

pparab
Genomic Map