Assay Details
| Gene Symbol: pparab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102567.1 | NP_001096037.1 | 6 - 7 | 1136 | 63 | |
| XM_009297698.3 | XP_009295973.1 | 7 - 8 | 1524 | 63 | ||
| GenBank mRNA | DQ017618.1 | - | 1 - 2 | 248 | 63 | |
| DQ839548.1 | - | 6 - 7 | 1136 | 63 | ||
More Information
Set Membership: |
|

pparab
Genomic Map