Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 22 - 23 | 3796 | 65 | |
| XM_009294421.3 | XP_009292696.1 | 22 - 23 | 3963 | 65 | ||
| XM_017352243.2 | XP_017207732.1 | 22 - 23 | 3967 | 65 | ||
| XM_021468075.1 | XP_021323750.1 | 22 - 23 | 4026 | 65 | ||
| XM_021468076.1 | XP_021323751.1 | 22 - 23 | 4027 | 65 | ||
| XM_021468077.1 | XP_021323752.1 | 22 - 23 | 4027 | 65 | ||
| XM_021468078.1 | XP_021323753.1 | 22 - 23 | 4028 | 65 | ||
| XM_021468079.1 | XP_021323754.1 | 22 - 23 | 4029 | 65 | ||
| XM_021468080.1 | XP_021323755.1 | 22 - 23 | 4030 | 65 | ||
| XM_021468081.1 | XP_021323756.1 | 22 - 23 | 4030 | 65 | ||
More Information
Set Membership: |
|

trioa
Genomic Map