Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 25 - 26 | 4080 | 95 | |
| XM_009294421.3 | XP_009292696.1 | 25 - 26 | 4247 | 95 | ||
| XM_017352243.2 | XP_017207732.1 | 25 - 26 | 4251 | 95 | ||
| XM_021468075.1 | XP_021323750.1 | 25 - 26 | 4310 | 95 | ||
| XM_021468076.1 | XP_021323751.1 | 25 - 26 | 4311 | 95 | ||
| XM_021468077.1 | XP_021323752.1 | 25 - 26 | 4311 | 95 | ||
| XM_021468078.1 | XP_021323753.1 | 25 - 26 | 4312 | 95 | ||
| XM_021468079.1 | XP_021323754.1 | 25 - 26 | 4313 | 95 | ||
| XM_021468080.1 | XP_021323755.1 | 25 - 26 | 4314 | 95 | ||
| XM_021468081.1 | XP_021323756.1 | 25 - 26 | 4314 | 95 | ||
More Information
Set Membership: |
|

trioa
Genomic Map