Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 40 - 41 | 6116 | 139 | |
| XM_009294421.3 | XP_009292696.1 | 40 - 41 | 6295 | 139 | ||
| XM_021468075.1 | XP_021323750.1 | 41 - 42 | 6388 | 139 | ||
| XM_021468076.1 | XP_021323751.1 | 41 - 42 | 6389 | 139 | ||
| XM_021468077.1 | XP_021323752.1 | 41 - 42 | 6389 | 139 | ||
| XM_021468078.1 | XP_021323753.1 | 41 - 42 | 6390 | 139 | ||
| XM_021468079.1 | XP_021323754.1 | 41 - 42 | 6391 | 139 | ||
More Information
Set Membership: |
|

trioa
Genomic Map