Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 49 - 50 | 7839 | 98 | |
| XM_009294421.3 | XP_009292696.1 | 49 - 50 | 8018 | 98 | ||
| XM_021468076.1 | XP_021323751.1 | 50 - 51 | 8112 | 98 | ||
More Information
Set Membership: |
|

trioa
Genomic Map