Assay Details
| Gene Symbol: trioa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104526.1 | NP_001097996.1 | 54 - 55 | 8379 | 90 | |
| XM_009294421.3 | XP_009292696.1 | 54 - 55 | 8558 | 90 | ||
| XM_021468075.1 | XP_021323750.1 | 55 - 56 | 8654 | 90 | ||
| XM_021468076.1 | XP_021323751.1 | 55 - 56 | 8652 | 90 | ||
| XM_021468077.1 | XP_021323752.1 | 55 - 56 | 8643 | 90 | ||
More Information
Set Membership: |
|

trioa
Genomic Map