Assay Details
| Gene Symbol: nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104943.1 | NP_001098413.1 | 9 - 10 | 1222 | 64 | |
| XM_021468499.1 | XP_021324174.1 | 7 - 8 | 1150 | 64 | ||
| GenBank mRNA | BC151962.1 | - | 9 - 10 | 1222 | 64 | |
More Information
Set Membership: |
|

nt5dc1
Genomic Map