Assay Details
| Gene Symbol: oscp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110123.1 | NP_001103593.1 | 3 - 4 | 410 | 109 | |
| XM_009293947.3 | XP_009292222.1 | 3 - 4 | 448 | 109 | ||
| GenBank mRNA | BC153926.1 | - | 3 - 4 | 410 | 109 | |
More Information
Set Membership: |
|

oscp1a
Genomic Map