Assay Details
| Gene Symbol: chico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312685.1 | NP_001299614.1 | 3 - 4 | 419 | 116 | |
| NM_130911.2 | NP_570986.1 | 2 - 3 | 668 | 116 | ||
| XM_009298917.3 | XP_009297192.1 | 2 - 3 | 708 | 116 | ||
| GenBank mRNA | AF085218.1 | - | 2 - 3 | 630 | 116 | |
| BC097097.1 | - | 3 - 4 | 419 | 116 | ||
More Information
Set Membership: |
|

chico
Genomic Map