Assay Details
| Gene Symbol: chico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312685.1 | NP_001299614.1 | 4 - 5 | 486 | 78 | |
| NM_130911.2 | NP_570986.1 | 3 - 4 | 735 | 78 | ||
| XM_009298917.3 | XP_009297192.1 | 3 - 4 | 775 | 78 | ||
| GenBank mRNA | AF085218.1 | - | 3 - 4 | 697 | 78 | |
| BC097097.1 | - | 4 - 5 | 486 | 78 | ||
More Information
Set Membership: |
|

chico
Genomic Map