Assay Details
| Gene Symbol: fzd8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130918.2 | NP_570993.2 | 1 | 1551 | 86 | |
| GenBank mRNA | AF039412.1 | - | 1 | 1504 | 86 | |
| AF117389.1 | - | 1 | 1237 | 86 | ||
| BC056273.1 | - | 1 | 1551 | 86 | ||
| BC063227.1 | - | 1 | 1549 | 86 | ||
More Information
Set Membership: |
|

fzd8a
Genomic Map