Assay Details
| Gene Symbol: fzd8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130918.2 | NP_570993.2 | 1 | 2149 | 147 | |
| GenBank mRNA | AF039412.1 | - | 1 | 2102 | 146 | |
| AF060697.1 | - | 1 | 2150 | 147 | ||
| BC056273.1 | - | 1 | 2149 | 147 | ||
| BC063227.1 | - | 1 | 2147 | 147 | ||
More Information
Set Membership: |
|

fzd8a
Genomic Map