Assay Details
| Gene Symbol: rho | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131084.1 | NP_571159.1 | 1 | 742 | 91 | |
| GenBank mRNA | AF105152.1 | - | 1 | 742 | 91 | |
| AF109368.1 | - | 1 | 744 | 91 | ||
| BC045288.1 | - | 1 | 734 | 91 | ||
| BC063938.1 | - | 1 | 709 | 91 | ||
| BC164171.1 | - | 1 | 677 | 91 | ||
| HM367063.1 | - | 1 | 643 | 91 | ||
| KT008392.1 | - | 1 | 643 | 91 | ||
| L11014.1 | - | 1 | 742 | 91 | ||
More Information
Set Membership: |
|

rho
Genomic Map