Assay Details
| Gene Symbol: atoh1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131091.2 | NP_571166.2 | 1 | 785 | 68 | |
| GenBank mRNA | AF024536.2 | - | 1 | 755 | 68 | |
| BC117598.1 | - | 1 | 777 | 68 | ||
| EH591361.1 | - | 1 | 803 | 68 | ||
| EH608855.1 | - | 1 | 848 | 68 | ||
More Information
Set Membership: |
|

atoh1a
Genomic Map