Assay Details
| Gene Symbol: pou5f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131112.1 | NP_571187.1 | 2 - 3 | 1071 | 83 | |
| XM_005161103.4 | XP_005161160.1 | 2 - 3 | 1491 | 83 | ||
| XM_005161104.4 | XP_005161161.1 | 2 - 3 | 1494 | 83 | ||
| GenBank mRNA | BC049295.1 | - | 2 - 3 | 1089 | 83 | |
| D28548.1 | - | 2 - 3 | 1054 | 83 | ||
| X84224.1 | - | 1071 | 83 | |||
More Information
Set Membership: |
|

pou5f3
Genomic Map